Key Documents
252-1903-OEM
Foxx Life Sciences
Foxx Life Sciences EZwaste® HPLC Solvent Waste System, Secondary Container
HPLC Solvent Waste Safety Tray Secondary Container, for 20L-60L Carboys REV A, 1/EA
Select a Size
1 EA
$106.64
$106.64
Available to ship onMarch 22, 2025Details
Select a Size
Change View
1 EA
$106.64
About This Item
PDF Icon
Shipped & Sold By Foxx Life Sciences
A Trusted Marketplace Seller
PDF Icon
Have a question?
$106.64
Available to ship onMarch 22, 2025Details
manufacturer/tradename
Foxx Life Sciences
General description
Foxx Life Sciences' Secondary Container is a perfect solution for preventing leakage and spillage from Foxx Life Sciences 1L, 2.5L, 4L, 5L, and 10L EZwaste® Solvent Waste System containers. Made from HDPE material, these containers are built to be tough and durable. Our Secondary Containers have the ability to hold at least 115% of the container's content.
Check our secondary containers' specifications to see if they fit your container.
Made to Order
Choose from one of the most recent versions:
Certificates of Analysis (COA)