LHAB-1000R-ST
Fluotics
Fluotics Automation Tips for Tecan Evo/Fluent Liha P1000 Pipet Tips
1000 uL, clear, refill with tip box, sterile, 96 format
Synonym(s):
Disposable pipette tips, refill, sterile, Evo, Fluent
About This Item
binding type
non-treated surface
volume
1000 μL
manufacturer/tradename
Fluotics
General description
Fluotics produces its Tecan Freedom EVO and Fluent compatible P1000 tips from the highest grade virgin polypropylene. We use ultra polishing techniques to ensure low liquid retention without the use of releasing agents or other additives. When using Fluotics automation tips a separate labware definition is unnecessary and existing automation control software protocols also do not need to be modified.
- Working volume: 10.00 - 1000.00 uL
- Tip format: sterile
- Well format: 96, 384 plate accessible
- Head: air and liquid LiHa and FCA Arm
- Tip material: polypropylene
- Tip box material: polypropylene
- Case quantity: refill with tip box base included, 96 tips per rack and 24 racks per case