ENAH9082208D
Enamine
1-(aminooxy)-3,3-dimethylbutan-2-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-(aminooxy)-3,3-dimethylbutan-2-ol
Inchi Code
1S/C6H15NO2/c1-6(2,3)5(8)4-9-7/h5,8H,4,7H2,1-3H3
InChI key
FSNWKKDILGLXFA-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Ice Pack
Physical Form
oil
Product Link
https://www.enaminestore.com/catalog/EN300-5252461
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-5252461
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.