Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH3035E313

Enamine

4-(2-methylbutan-2-yl)cyclohexane-1-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
179.31
MDL number:
Product ENAH3035E313 is currently not available for purchase. Contact Technical Service

Iupac Name

4-tert-pentylcyclohexanecarbonitrile

Inchi Code

1S/C12H21N/c1-4-12(2,3)11-7-5-10(9-13)6-8-11/h10-11H,4-8H2,1-3H3

InChI key

YGOQJPOQECSGQK-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-6493761

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-6493761


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library