Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH3035D09F

Enamine

2-(quinolin-6-yl)acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
168.2
MDL number:
Product ENAH3035D09F is currently not available for purchase. Contact Technical Service

mp

80-82

Iupac Name

6-quinolinylacetonitrile

Inchi Code

1S/C11H8N2/c12-6-5-9-3-4-11-10(8-9)2-1-7-13-11/h1-4,7-8H,5H2

InChI key

SGSMUSDVKCMDGR-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-312337

Melting Point

80-82

MSDS

https://www.enaminestore.com/catalog/EN300-312337


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library