Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH0CD45A47

Enamine

1-propyl-1H-imidazole-5-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.17
MDL number:
Product ENAH0CD45A47 is currently not available for purchase. Contact Technical Service

Iupac Name

1-propyl-1H-imidazole-5-carbaldehyde

Inchi Code

1S/C7H10N2O/c1-2-3-9-6-8-4-7(9)5-10/h4-6H,2-3H2,1H3

InChI key

CZNHVLOXIAQFJC-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-95410

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-95410


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library