Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENAH03A1A0DC

Enamine

2-bromo-4-methyl-1-(pentafluoroethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
289.04
MDL number:
Product ENAH03A1A0DC is currently not available for purchase. Contact Technical Service

Iupac Name

2-bromo-4-methyl-1-(perfluoroethyl)benzene

Inchi Code

1S/C9H6BrF5/c1-5-2-3-6(7(10)4-5)8(11,12)9(13,14)15/h2-4H,1H3

InChI key

HNMJLGNQJDODQS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-1198034

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-1198034


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library