Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA580348211

Enamine

3-(2-chloropyridin-3-yl)propan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
171.63
MDL number:

Iupac Name

3-(2-chloropyridin-3-yl)propan-1-ol

Inchi Code

1S/C8H10ClNO/c9-8-7(4-2-6-11)3-1-5-10-8/h1,3,5,11H,2,4,6H2

InChI key

JCOOCSQBENUWFS-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-311297

MSDS

https://www.enaminestore.com/catalog/EN300-311297


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library