Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA508626256

Enamine

3-cyclopropyl-4-methylpyridin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
148.21
MDL number:
Product ENA508626256 is currently not available for purchase. Contact Technical Service

Iupac Name

3-cyclopropyl-4-methyl-2-pyridinamine

Inchi Code

1S/C9H12N2/c1-6-4-5-11-9(10)8(6)7-2-3-7/h4-5,7H,2-3H2,1H3,(H2,10,11)

InChI key

YQWOAHJDBWFKIT-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-295124

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-295124


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library