Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA458430163

Enamine

methyl 4-(2,2,2-trifluoroethoxy)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
234.17
MDL number:
Product ENA458430163 is currently not available for purchase. Contact Technical Service

mp

55-57

Iupac Name

methyl 4-(2,2,2-trifluoroethoxy)benzoate

Inchi Code

1S/C10H9F3O3/c1-15-9(14)7-2-4-8(5-3-7)16-6-10(11,12)13/h2-5H,6H2,1H3

Inchi Key

IJQLWPBDYFRONS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-230953

Melting Point

55-57

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-230953


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library