Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA447957502

Enamine

6-bromoquinoline-8-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
306.57
MDL number:

Iupac Name

6-bromo-8-quinolinesulfonyl chloride

Inchi Code

1S/C9H5BrClNO2S/c10-7-4-6-2-1-3-12-9(6)8(5-7)15(11,13)14/h1-5H

InChI key

GKHOYDOQDYWDGB-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-207279

MSDS

https://www.enaminestore.com/catalog/EN300-207279


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library