Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA429359058

Enamine

ethyl 2-(5-aminopyridin-2-yl)acetate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
180.21
MDL number:
Product ENA429359058 is currently not available for purchase. Contact Technical Service

Iupac Name

ethyl (5-amino-2-pyridinyl)acetate

Inchi Code

1S/C9H12N2O2/c1-2-13-9(12)5-8-4-3-7(10)6-11-8/h3-4,6H,2,5,10H2,1H3

InChI key

PXKDDIRASPIURS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

oil

Product Link

https://www.enaminestore.com/catalog/EN300-186922

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-186922


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library