Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA428074168

Enamine

4,5-dichlorothiophene-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
178.04
MDL number:

Iupac Name

4,5-dichlorothiophene-3-carbonitrile

Inchi Code

1S/C5HCl2NS/c6-4-3(1-8)2-9-5(4)7/h2H

InChI key

GLUMTSGBCKJDHN-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-180868

pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.enaminestore.com/catalog/EN300-180868


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library