Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA410542943

Enamine

(6-methoxy-1,2,3,4-tetrahydronaphthalen-1-yl)methanamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
191.27
MDL number:

mp

190-192

Iupac Name

(6-methoxy-1,2,3,4-tetrahydro-1-naphthalenyl)methanamine

Inchi Code

1S/C12H17NO/c1-14-11-5-6-12-9(7-11)3-2-4-10(12)8-13/h5-7,10H,2-4,8,13H2,1H3

InChI key

PWCAWOYQXQCEEV-UHFFFAOYSA-N

storage temp.

4

Purity

94

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-138590

Melting Point

190-192

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-138590


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library