Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA390552662

Enamine

{3-fluoro-4-[(1H-pyrazol-1-yl)methyl]phenyl}boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
220.01
MDL number:

Iupac Name

3-fluoro-4-(1H-pyrazol-1-ylmethyl)phenylboronic acid

Inchi Code

1S/C10H10BFN2O2/c12-10-6-9(11(15)16)3-2-8(10)7-14-5-1-4-13-14/h1-6,15-16H,7H2

InChI key

FDVOYBMPDRQBGK-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-127494

MSDS

https://www.enaminestore.com/catalog/EN300-127494


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library