Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA390347431

Enamine

methyl[1-(naphthalen-1-yl)ethyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
185.27
MDL number:
Product ENA390347431 is currently not available for purchase. Contact Technical Service

Iupac Name

N-methyl-1-(1-naphthyl)ethanamine

Inchi Code

1S/C13H15N/c1-10(14-2)12-9-5-7-11-6-3-4-8-13(11)12/h3-10,14H,1-2H3

InChI key

YYETYHLXPGYQPZ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-126295

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-126295


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library