Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA369383576

Enamine

4-bromo-2-nitrobenzene-1-thiol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
234.07
MDL number:

Iupac Name

4-bromo-2-nitrobenzenethiol

Inchi Code

1S/C6H4BrNO2S/c7-4-1-2-6(11)5(3-4)8(9)10/h1-3,11H

InChI key

AJQYWSYQGXVQBM-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-99775

pictograms

CorrosionExclamation mark

Signal Word

Danger

MSDS

https://www.enaminestore.com/catalog/EN300-99775


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library