Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA369383166

Enamine

2-{[(furan-2-yl)methyl]amino}benzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
198.22
MDL number:

Iupac Name

2-[(2-furylmethyl)amino]benzonitrile

Inchi Code

1S/C12H10N2O/c13-8-10-4-1-2-6-12(10)14-9-11-5-3-7-15-11/h1-7,14H,9H2

InChI key

KUDSLEWSBLTGCG-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-32906

MSDS

https://www.enaminestore.com/catalog/EN300-32906


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library