Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA238711260

Enamine

2-(2,6-difluorophenyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
156.13
MDL number:
Product ENA238711260 is currently not available for purchase. Contact Technical Service

Iupac Name

2-(2,6-difluorophenyl)oxirane

Inchi Code

1S/C8H6F2O/c9-5-2-1-3-6(10)8(5)7-4-11-7/h1-3,7H,4H2

InChI key

HKEQMAWILPCWGK-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-44844

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library