Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA048303627

Enamine

2-chloro-N-{[(4-chlorophenyl)carbamoyl]methyl}-N-methylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
275.13
MDL number:

Iupac Name

2-chloro-N-[2-(4-chloroanilino)-2-oxoethyl]-N-methylacetamide

Inchi Code

1S/C11H12Cl2N2O2/c1-15(11(17)6-12)7-10(16)14-9-4-2-8(13)3-5-9/h2-5H,6-7H2,1H3,(H,14,16)

InChI key

SWDWMDRPDBBGDL-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-11700

MSDS

https://www.enaminestore.com/catalog/EN300-11700


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library