Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ENA018108561

Enamine

5-cyclopropyl-1-phenyl-1H-1,2,4-triazole-3-thiol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
217.29
MDL number:
Product ENA018108561 is currently not available for purchase. Contact Technical Service

Iupac Name

5-cyclopropyl-1-phenyl-1H-1,2,4-triazol-3-yl hydrosulfide

Inchi Code

1S/C11H11N3S/c15-11-12-10(8-6-7-8)14(13-11)9-4-2-1-3-5-9/h1-5,8H,6-7H2,(H,13,15)

InChI key

YLIGLBKYJFKWOD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-05263

MSDS

https://www.enaminestore.com/catalog/EN300-05263


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library