Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PH0786E

Eisco Labs

Eisco Labs Bar Magnets

Bar Magnets - Chrome Steel, 150x12x5mm

Synonym(s):

Bar Magnets, Chrome Magnets, Steel Magnets

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$9.99

$9.99


Available to ship onFebruary 25, 2025Details



Select a Size

Change View
1 EA
$9.99

About This Item

PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?

$9.99


Available to ship onFebruary 25, 2025Details


manufacturer/tradename

Eisco Labs

General description

  • Chrome steel magnets
  • Perform a variety of activities and experiments involving magnetism
  • Low magnetic stregnth for careful experiments

Includes: (1) Pair of Magnets

Economy bar magnets with keepers.

Perform a variety of activities and experiments involving magnetism.

The magnet keeper helps preserve the strength of the magnet by completing the magnetic circuit, and is particularly useful for economy magnets like these. Keepers also have a useful safety function as they stop external metal being attracted to the magnet.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library