Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PH0064D

Eisco Labs

Eisco Labs Half Meter Stick, Hardwood

Half Meter Stick, Hardwood 50cm

Synonym(s):

Meter Stick, Measuring Stick, Wooden Measuring Stick

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
Product PH0064D is currently not available for purchase. Contact Eisco Labs.

manufacturer/tradename

Eisco Labs

General description

  • Hardwood half meter stick (50cm)
  • Graduated in both centimeters (cm) and millimeters (mm)
  • Ruler Overall Dimensions: 50cm x 2.5cm x 0.6cm
  • Zero top
  • 50cm Hard Wood Half Meter Stick, Graduated in Centimeters and Millimeters


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library