CHWB1015PK2
Eisco Labs
Eisco Labs LDPE Wash Bottle for Acetone
2PK LDPE Wash Bottle for Acetone, 1000ml (4 Colors) - Wide Mouth
Synonym(s):
Wash Bottle, Wide Mouth, Wide Mouth Wash Bottle, Squeeze Bottle
About This Item
manufacturer/tradename
Eisco Labs
General description
- Two pack Wash Bottles, 1000ml capacity each
- Easy to read labeling, including chemical information and safety warnings (4 colors)
- Self venting design
- Made of low density polyethylene
- Color coded screw cap for easy identification (4 colors)
Performance plastic with flex ink. Ink won't fade, crack or peel. Each bottle features a two piece construction, consisting of the bottle and leak proof cap with built in nozzle. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
Pack of 2 wash bottles featuring easy to read labeling and safety warnings. Color code labeled for Acetone (4 colors), with screw cap and nozzle. 1000ml capacity. Self venting.
Each bottle measures 10.75" in total height (7" bottle alone) and 3.5" diameter. Bottle opening has an inner diameter of 1.7" and an outer diameter measuring 1.9".
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.