Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH0652BR8

Eisco Labs

Eisco Labs Rectangular Retort Stand, Rod & Ring Set

5 Piece Set - Rectangular Retort Stand, Rod & Ring Set

Synonym(s):

Stand, Retort Stand, Rods, Rings

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$67.99

$67.99


Available to ship onFebruary 18, 2025Details



Select a Size

Change View
1 EA
$67.99

About This Item

PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?

$67.99


Available to ship onFebruary 18, 2025Details


manufacturer/tradename

Eisco Labs

General description

  • HIGH QUALITY || This laboratory set includes retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10mm x 1.5mm metric thread and three cast iron support rings
  • STABILITY || The 8" long by 5" wide base weighs 1.75lbs, providing superb stability for experiments and setups. The included rod measures 23.6" tall and is 12mm in diameter. Includes three support rings, with diameters of 3", 4" and 5" to support a variety of apparatus
  • TAPPED HOLE AND THREADED ROD || Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10mm x 1.5mm metric thread
  • GREAT FOR CLASSROOM and HOME LABORATORIES || The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories and basic professional laboratory setups
  • MANUFACTURED BY EISCO LABS || Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind

5 Piece Laboratory Set || Retort Base, 8" x 5" - Stainless Steel Rod, 23.6" - 3 Support Rings (3", 4" and 5")

Lab stands are used in every area of science - especially chemistry, for holding and manipulating an experimental apparatus. If you want to create a home laboratory, a retort base and rod are essential for any experiment setup. This set includes an assortment of cast iron rings to support a variety of laboratory apparatus.

The retort stand is made of corrosion and scaling resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10mm x 1.5mm metric thread. Stand measures 8" long, 5" wide and is approximately 1" in height. Included rod is made of stainless steel and measures 23.6" tall, 12mm in diameter and features a 10x1.5mm metric thread. Three cast iron support rings with diameters of 3", 4" and 5". Each ring features a zinc plated steel shank secured to a screw clamp. Distance from each of the rings centers to the center of the support measures 4.75".

Stand weighs 1.75lbs, providing superb stability for basic experiments and setups. The durable and sturdy construction of this set makes it an excellent choice for science classrooms, household laboratories and basic professional laboratories.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library