Key Documents
CH0520
Eisco Labs
Eisco Labs Deflagrating Spoon & Cap
Deflagrating Spoon & Cap - Steel Cup & Rod, Aluminum Cap
Synonym(s):
Deflagrating Spoon & Cap, Spoon and Cap, Cap, Spoon
Select a Size
1 EA
$9.99
$9.99
Available to ship onFebruary 04, 2025Details
Select a Size
Change View
1 EA
$9.99
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
$9.99
Available to ship onFebruary 04, 2025Details
manufacturer/tradename
Eisco Labs
General description
  • Spoon measures approximately 35 cm in length, and caps measure approximately 90 mm in diameter
  • Spoon bowl measures approximately 15 mm in diameter
  • Long spoon and wide cap protect user from extreme exothermic reactions
  • Great in both professional and educational lab settings
These spoons are often used for burning sulfur, phosphorus, and other heavily reactive materials.
Deflagrating spoons are perfect for handling strong exothermic reactions such as these, allowing the user to move the material while keeping a safe distance. The cap attaches to the spoon and provides another layer of safety.
Choose from one of the most recent versions:
Certificates of Analysis (COA)