Key Documents
CH0430A
Eisco Labs
Eisco Labs Glass Conical Erlenmeyer Flask With PTFE Screw Cap
Erlenmeyer Flask, 25ml - Borosilicate Glass - With PTFE Screw Cap - Conical Shape - White Graduations
Synonym(s):
Erlenmeyer Flask, Flask, Glass Flask
Select a Size
1 EA
$11.99
$11.99
Available to ship onFebruary 04, 2025Details
Select a Size
Change View
1 EA
$11.99
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
$11.99
Available to ship onFebruary 04, 2025Details
manufacturer/tradename
Eisco Labs
General description
  • The base diameter of the bottle is 1.8" and the overall height is 3.7". Please note: these are hand blown with the capacity in mind and overall dimensions can vary slightly.
  • Hand blown premium borosilicate 3.3 glass.
  • The cap is made of polypropylene, has a PTFE liner, and is acid proof and autoclavable.
  • Designed to store liquids in the laboratory, but can be used in the home as well -- or even in a set for "potions".
  • Top rack dishwasher safe and autoclavable.
Premium borosilicate Erlenmeyer flasks with acid proof screw caps. Eisco glassware is designed to be used in labs but is often used in homes or garages as well.
Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)