Key Documents
CH0424E
Eisco Labs
Eisco Labs Glass Conical Erlenmeyer Flask with Narrow Neck
Erlenmeyer Flask, 150ml - Borosilicate Glass - Narrow Neck, Conical Shape - White Graduations
Synonym(s):
Erlenmeyer Flask, Flask, Glass Flask
Select a Size
1 EA
$12.99
$12.99
Available to ship onFebruary 25, 2025Details
Select a Size
Change View
1 EA
$12.99
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
$12.99
Available to ship onFebruary 25, 2025Details
manufacturer/tradename
Eisco Labs
General description
  • High Quality Borosilicate glass
  • 150ml with 25ml graduations
  • (1) Single Flask
1 Single Erlenmeyer conical Flask 150ml, narrow neck, Made from high quality borosilicate glass. 150ml with 25ml graduations
Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)