Key Documents
CH0268E
Eisco Labs
Eisco Labs Glass Centrifuge Tube
50ml
Synonym(s):
Centrifuge Tube, Tubing, Glass Tube
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Eisco Labs
General description
  • Made of high quality, heavy duty borosilicate glass
  • 50ml capacity
  • Conical shape, beaded rim
  • Measures 5.25" tall and has an outer diameter of 1.1"
  • Excellent for science classrooms, professional laboratories and home use
50ml capacity centrifuge tube made of high quality, heavy duty Borosilicate 3.3 glass. Features conical shape and a beaded rim.
Tube measures 5.25" tall, and has an inner diameter measuring 1", and an outer diameter measuring 1.1".
Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures & thermal shocks. Eisco Labs Borosilicate glass has the following heat tolerances: Strain Point -515°C | Annealing Point 565°C | Softening Point 820°C.
Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.