Key Documents
BTAUT50
Eisco Labs
Eisco Labs Burette
Automatic, Self-Zeroing, Self-Supporting, Closed System, Borosilicate 50ml Burette with Reservoir, Tube, and Base
Synonym(s):
Self Zero, Glass Burette, Auto Burette
Select a Size
1 EA
$187.99
$187.99
Available to ship onMarch 04, 2025Details
Select a Size
Change View
1 EA
$187.99
About This Item
PDF Icon
Shipped & Sold By Eisco Labs
A Trusted Marketplace Seller
PDF Icon
Have a question?
$187.99
Available to ship onMarch 04, 2025Details
manufacturer/tradename
Eisco Labs
General description
  • 50ML / 500ML || Automatic zero burette with 50ml capacity mounted on a 2000ml reservoir - Includes automatic zero facility - 0.10ml sub. divisions
  • FEATURES || Self-supporting base - Polyethylene filling tube to connect burette to reservoir - Flexible burette tip - Reservoir screw cap with fitting for polyethylene tube
  • STOPCOCK || Features an automatic, push button-style stopcock which facilitates the complete drainage of excess liquid back into the reservoir
  • DURABLE & STURDY || Burette is made of precisely bore high quality, heavy duty Borosilicate 3.3 glass - Built to last
  • PREMIUM QUALITY || Features chemical resistant blue printing - All contents are included in a single package
Automatic 50ml burette made of boroscilicate glass which is self-zeroing. 0.1ml resolution with permanent blue markings against a white background for easy viewing. Stopcock is pushbutton style for ease of use to deliver fluids drop by drop. Attached to the stop cock is a screw clamp for repeat titrations. This set up is a complete self supporting structure with a heavy duty base made of polyethylene which the reservoir snaps into. The advantage to a system such as this is that it is a completely closed system. Package includes: -50ml automatic burette -Automatic stop cock with screw setting and release -500ml reservoir -Self Supporting base -Polyethylene filling Tube to connect burette to reservoir -Flexible burette tip -Reservoir screw cap with fitting for polyethylene tube Calibrated for delivery. Eisco Labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Choose from one of the most recent versions:
Certificates of Analysis (COA)