Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH2F649E36

Combi-Blocks, Inc.

(1,3-Dioxoisoindol-2-yl)(4-fluorophenyl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
299.26
MDL number:
Product COMH2F649E36 is currently not available for purchase. Contact Technical Service

Iupac Name

2-(1,3-dioxoisoindolin-2-yl)-2-(4-fluorophenyl)acetic acid

Inchi Code

1S/C16H10FNO4/c17-10-7-5-9(6-8-10)13(16(21)22)18-14(19)11-3-1-2-4-12(11)15(18)20/h1-8,13H,(H,21,22)

InChI key

WBZCVCORTWFCIF-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?SH-7134

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?SH-7134


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library