Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH04239836

Combi-Blocks, Inc.

1-(Thien-2-ylmethyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
182.29
MDL number:
Product COMH04239836 is currently not available for purchase. Contact Technical Service

Iupac Name

1-(2-thienylmethyl)piperazine

Inchi Code

1S/C9H14N2S/c1-2-9(12-7-1)8-11-5-3-10-4-6-11/h1-2,7,10H,3-6,8H2

InChI key

XTAPGYLDPZMKRN-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?ST-9347

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?ST-9347


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library