Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH04238929

Combi-Blocks, Inc.

N-Methyl-2-morpholin-4-ylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
158.2
MDL number:
Product COMH04238929 is currently not available for purchase. Contact Technical Service

Iupac Name

N-methyl-2-morpholinoacetamide

Inchi Code

1S/C7H14N2O2/c1-8-7(10)6-9-2-4-11-5-3-9/h2-6H2,1H3,(H,8,10)

InChI key

UBGWYKSEGWKKFU-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6333

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-6333


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library