Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH04237E2E

Combi-Blocks, Inc.

N-Allyl-4-oxo-3,4-dihydrophthalazine-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
229.24
MDL number:
Product COMH04237E2E is currently not available for purchase. Contact Technical Service

Iupac Name

N-allyl-4-oxo-3,4-dihydro-1-phthalazinecarboxamide

Inchi Code

1S/C12H11N3O2/c1-2-7-13-12(17)10-8-5-3-4-6-9(8)11(16)15-14-10/h2-6H,1,7H2,(H,13,17)(H,15,16)

InChI key

RHZKAEYCYUEAML-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3485

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-3485


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library