Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COMH04235B61

Combi-Blocks, Inc.

5-Nitro-2-(1h-pyrrol-1-yl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
204.19
MDL number:

Iupac Name

5-nitro-2-(1H-pyrrol-1-yl)phenol

Inchi Code

1S/C10H8N2O3/c13-10-7-8(12(14)15)3-4-9(10)11-5-1-2-6-11/h1-7,13H

InChI key

DZDJALIVRCUNIY-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QY-4528

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QY-4528


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library