Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM517298064

Combi-Blocks, Inc.

3-(5-Fluoropyridin-2-yloxy)aniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
204.2
MDL number:

Iupac Name

3-[(5-fluoro-2-pyridinyl)oxy]phenylamine

Inchi Code

1S/C11H9FN2O/c12-8-4-5-11(14-7-8)15-10-3-1-2-9(13)6-10/h1-7H,13H2

InChI key

WOFXPGGXIADBOP-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QY-2277

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QY-2277


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library