Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM490020580

Combi-Blocks, Inc.

2,2,2-trifluoro-N-(4-fluorophenyl)Ethanimidoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.57
MDL number:
Product COM490020580 is currently not available for purchase. Contact Technical Service

Iupac Name

(E)-2,2,2-trifluoro-N-(4-fluorophenyl)acetimidoyl chloride

Inchi Code

1S/C8H4ClF4N/c9-7(8(11,12)13)14-6-3-1-5(10)2-4-6/h1-4H/b14-7+

InChI key

RQMRLWLPPFJICW-VGOFMYFVSA-N

storage temp.

Refrigerated

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?OT-4158

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?OT-4158


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library