Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448685408

Combi-Blocks, Inc.

2-Fluoro-5-(3-hydroxyphenyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
232.21
MDL number:

Iupac Name

4-fluoro-3'-hydroxy[1,1'-biphenyl]-3-carboxylic acid

Inchi Code

1S/C13H9FO3/c14-12-5-4-9(7-11(12)13(16)17)8-2-1-3-10(15)6-8/h1-7,15H,(H,16,17)

InChI key

KBMWODWZENMSCQ-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-8933

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-8933


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library