Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448685298

Combi-Blocks, Inc.

2-(4-t-butylphenyl)Isonicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.32
MDL number:
Product COM448685298 is currently not available for purchase. Contact Technical Service

Iupac Name

2-(4-tert-butylphenyl)isonicotinic acid

Inchi Code

1S/C16H17NO2/c1-16(2,3)13-6-4-11(5-7-13)14-10-12(15(18)19)8-9-17-14/h4-10H,1-3H3,(H,18,19)

InChI key

YOVPEGIHXKWZTA-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-7400

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-7400


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library