COM448685081
Combi-Blocks, Inc.
2-(3-BOC-Aminophenyl)phenol
About This Item
Iupac Name
3-[(tert-butoxycarbonyl)amino]-2'-hydroxy-1,1'-biphenyl
Inchi Code
1S/C17H19NO3/c1-17(2,3)21-16(20)18-13-8-6-7-12(11-13)14-9-4-5-10-15(14)19/h4-11,19H,1-3H3,(H,18,20)
Inchi Key
HYJWABFIDNHSGT-UHFFFAOYSA-N
Storage temp.
Purity
Country of Origin
Product Link
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?YA-1897
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.