COM448665924
Combi-Blocks, Inc.
3-Ethoxycarbonyl-5,6-dihydro-2-methyl-4H-pyran
About This Item
Iupac Name
ethyl 6-methyl-3,4-dihydro-2H-pyran-5-carboxylate
Inchi Code
1S/C9H14O3/c1-3-11-9(10)8-5-4-6-12-7(8)2/h3-6H2,1-2H3
InChI key
BWVSWJMSYLYIJA-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Product Link
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QE-3967
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.