COM448635226
Combi-Blocks, Inc.
5-Chloro-3-ethoxy-2-methoxypyridine
Sign Into View Organizational & Contract Pricing
Select a Size
All Photos(1)
Select a Size
Change View
About This Item
Iupac Name
5-chloro-3-ethoxy-2-methoxypyridine
Inchi Code
1S/C8H10ClNO2/c1-3-12-7-4-6(9)5-10-8(7)11-2/h4-5H,3H2,1-2H3
InChI key
BLNYKLDTGCZLRJ-UHFFFAOYSA-N
Purity
96%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?PY-1403
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?PY-1403
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.