Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448621201

Combi-Blocks, Inc.

1-Ethoxymethyl-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
126.16
MDL number:
Product COM448621201 is currently not available for purchase. Contact Technical Service

Iupac Name

1-(ethoxymethyl)-1H-imidazole

Inchi Code

1S/C6H10N2O/c1-2-9-6-8-4-3-7-5-8/h3-5H,2,6H2,1H3

InChI key

VAJJXKGFPILYBO-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-1021

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HI-1021


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library