Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COM448614780

Combi-Blocks, Inc.

3-Amino-5-(morpholinocarbonyl)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
250.06
MDL number:

Iupac Name

3-amino-5-(4-morpholinylcarbonyl)phenylboronic acid

Inchi Code

1S/C11H15BN2O4/c13-10-6-8(5-9(7-10)12(16)17)11(15)14-1-3-18-4-2-14/h5-7,16-17H,1-4,13H2

InChI key

CKXOOFBRAGWZHT-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4430

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-4430


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library