CIAH98DE39DF
ChemScene LLC (Preferred Partner)
tert-Butyl (1-(methylsulfonyl)piperidin-3-yl)carbamate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
tert-butyl (1-(methylsulfonyl)piperidin-3-yl)carbamate
Inchi Code
1S/C11H22N2O4S/c1-11(2,3)17-10(14)12-9-6-5-7-13(8-9)18(4,15)16/h9H,5-8H2,1-4H3,(H,12,14)
InChI key
UXYFDFIFACOEAI-UHFFFAOYSA-N
storage temp.
Store at room temperature
Purity
95%
Country of Origin
CN
Physical Form
Solid
Product Link
https://www.chemscene.com/1262310-00-8.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0618478.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.