Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH9884F414

ChemScene LLC (Preferred Partner)

2-(1-Oxo-1,2-dihydroisoquinolin-3-yl)acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
203.2
MDL number:
Product CIAH9884F414 is currently not available for purchase. Contact Technical Service

bp

518.7+-50.0 C at 760 mmHg

Iupac Name

2-(1-hydroxyisoquinolin-3-yl)acetic acid

Inchi Code

1S/C11H9NO3/c13-10(14)6-8-5-7-3-1-2-4-9(7)11(15)12-8/h1-5H,6H2,(H,12,15)(H,13,14)

InChI key

SXFIXVKLRFTKOH-UHFFFAOYSA-N

storage temp.

-20C, sealed storage, away from moisture

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/69391-35-1.html

Boiling point

518.7+-50.0 C at 760 mmHg

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0527267.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library