Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CIAH987EC857

ChemScene LLC (Preferred Partner)

5-Bromo-2-cyclopropoxypyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
214.06
MDL number:

Iupac Name

5-bromo-2-(cyclopropyloxy)pyridine

Inchi Code

1S/C8H8BrNO/c9-6-1-4-8(10-5-6)11-7-2-3-7/h1,4-5,7H,2-3H2

InChI key

WFGHZOZHSRYNKT-UHFFFAOYSA-N

storage temp.

Store at room temperature

Purity

98%

Country of Origin

CN

Physical Form

Liquid

Product Link

https://www.chemscene.com/1243318-10-6.html

Signal Word

Warning

Hazard Statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0097659.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library