Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C12H316CEB7C

Chemcia Scientific, LLC

3-Bromo-2H-indazole-6-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
255.07
MDL number:
Product C12H316CEB7C is currently not available for purchase. Contact Technical Service

Iupac Name

methyl 3-bromo-2H-indazole-6-carboxylate

Inchi Code

1S/C9H7BrN2O2/c1-14-9(13)5-2-3-6-7(4-5)11-12-8(6)10/h2-4H,1H3,(H,11,12)

InChI key

NXMJEXMODINMIJ-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

White Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=3469

MSDS

http://www.chemcia.com/product_fl02.asp?id=3469


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library