Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T-067

Supelco

Tapentadol-D3-β-D-glucuronide solution

100 μg/mL in acetonitrile: water, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D3H28NO7
Molecular Weight:
400.48
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in acetonitrile: water

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])N(C)C[C@H](C)[C@@H](CC)c1cccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(O)=O)c1

InChI

1S/C20H31NO7/c1-5-14(11(2)10-21(3)4)12-7-6-8-13(9-12)27-20-17(24)15(22)16(23)18(28-20)19(25)26/h6-9,11,14-18,20,22-24H,5,10H2,1-4H3,(H,25,26)/t11-,14+,15-,16-,17+,18-,20+/m0/s1/i3D3

InChI key

CTYJDHSTNLOUMT-UTOPTPDDSA-N

General description

Tapentadol-ß-D-glucuronide is the primary urinary metabolite of tapentadol, the first new drug of the centrally-acting analgesic class approved in the US for pain management in more than 25 years. This new internal standard is suitable for many LC/MS or GC/MS applications including pain prescription monitoring, forensic testing, clinical toxicology, urine drug testing or isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service