Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SCT-022

Supelco

2,4,6-Trinitrotoluene

vial of 100 mg, analytical standard

Synonym(s):

TNT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5N3O6
CAS Number:
Molecular Weight:
227.13
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

packaging

vial of 100 mg

application(s)

forensics and toxicology

storage temp.

−20°C

SMILES string

Cc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Expl. 1.1 - STOT RE 2

Storage Class

1 - Explosive hazardous materials

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tommaso Rossi et al.
Investigative ophthalmology & visual science, 53(13), 8057-8066 (2012-11-01)
Primary blast injury (PBI) mostly affects air-filled organs, although it is sporadically reported in fluid-filled organs, including the eye. The purpose of the present paper is to explain orbit blast injury mechanisms through finite element modeling (FEM). FEM meshes of
Craig A McFarland et al.
Ecotoxicology (London, England), 21(8), 2372-2390 (2012-09-15)
Evaluation of multiple-stressor effects stemming from habitat degradation, climate change, and exposure to chemical contaminants is crucial for addressing challenges to ecological and environmental health. To assess the effects of multiple stressors in an understudied taxon, the western fence lizard
Liangdong Zou et al.
Applied biochemistry and biotechnology, 168(7), 1976-1988 (2012-10-19)
2,4,6-Trinitrotoluene (TNT), an extensively used and versatile explosive, is harmful in soil and water. In the present study, four bacterial strains capable of degrading TNT have been isolated from contaminated sites and named as Thu-A, Thu-B, Thu-C, and Thu-Z. Thu-Z
Ronit Freeman et al.
Advanced materials (Deerfield Beach, Fla.), 24(48), 6416-6421 (2012-09-26)
Chemically modified CdSe/ZnS quantum dots (QDs) are used as fluorescent probes for the analysis of explosives, and specifically, the detection of trinitrotoluene (TNT) or trinitrotriazine (RDX). The QDs are functionalized with electron-donating ligands that bind nitro-containing explosives, exhibiting electron-acceptor properties
Dongyue Lin et al.
Analytica chimica acta, 744, 92-98 (2012-09-01)
This study found that 1,2-ethylenediamine (EDA) as a primary amine could be modified onto the surface of citrate-stabilized gold nanoparticles (Au NPs), and the EDA-capped Au NPs were successfully used as an ultrasensitive optical probe for TNT detection. The strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service